Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018055816.1 | XP_017911305.1 | 8 - 9 | 1120 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |