Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005687432.3 | XP_005687489.1 | 3 - 4 | 801 | 65 | |
XM_018056765.1 | XP_017912254.1 | 3 - 4 | 385 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Capra hircus Probe spans exons |