Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005698825.3 | XP_005698882.2 | 2 - 3 | 304 | 68 | |
GenBank mRNA | KA319673.1 | - | 2 - 3 | 313 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Capra hircus Probe spans exons |