Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPTY2D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005699524.3 | XP_005699581.2 | 2 - 3 | 379 | 97 | |
GenBank mRNA | JO595947.2 | - | 4 - 5 | 1932 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |