Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPM1N | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018062605.1 | XP_017918094.1 | 3 - 4 | 1756 | 86 | |
GenBank mRNA | KA306823.1 | - | 3 - 4 | 257 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |