Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPM1N | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018062605.1 | XP_017918094.1 | 4 - 5 | 1816 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |