Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018061190.1 | XP_017916679.1 | 1 - 2 | 3862 | 73 | |
XM_018061191.1 | XP_017916680.1 | 1 - 2 | 1376 | 73 | ||
GenBank mRNA | KA339148.1 | - | 5 - 6 | 947 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |