Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFRSF13B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013971985.2 | XP_013827439.2 | 1 - 2 | 275 | 104 | |
XM_018064822.1 | XP_017920311.1 | 1 - 2 | 275 | 104 | ||
GenBank mRNA | JO612465.2 | - | 1 - 2 | 168 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |