Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CBX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018063651.1 | XP_017919140.1 | 5 | 761 | 86 | |
GenBank mRNA | KA330489.1 | - | 5 | 692 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Capra hircus Both primers and probe map within a single exon |