Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFI44L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005678249.3 | XP_005678306.3 | 5 - 6 | 1009 | 100 | |
GenBank mRNA | JO610806.2 | - | 4 - 5 | 609 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |