Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005682081.3 | XP_005682138.1 | 4 - 5 | 356 | 70 | |
GenBank mRNA | JO631683.2 | - | 1 - 2 | 396 | 70 | |
JT634338.1 | - | 1 - 2 | 183 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Capra hircus Probe spans exons |