Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM83F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018048797.1 | XP_017904286.1 | 3 - 4 | 815 | 79 | |
GenBank mRNA | JT662704.1 | - | 4 - 5 | 723 | 79 | |
KA346728.1 | - | 3 - 4 | 817 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |