Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLAC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018042301.1 | XP_017897790.1 | 2 - 3 | 264 | 79 | |
GenBank mRNA | KA326093.1 | - | 2 - 3 | 626 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Capra hircus Probe spans exons |