Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CREG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677108.3 | XP_005677165.3 | 3 - 4 | 717 | 105 | |
GenBank mRNA | JO511845.2 | - | 3 - 4 | 720 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Capra hircus Probe spans exons |