Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPAG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005683853.3 | XP_005683910.1 | 5 - 6 | 1905 | 96 | |
XM_018052154.1 | XP_017907643.1 | 5 - 6 | 1905 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Capra hircus Probe spans exons |