Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DOK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005683968.3 | XP_005684025.2 | 3 - 4 | 539 | 74 | |
GenBank mRNA | JO595694.2 | - | 3 - 4 | 580 | 74 | |
JO595696.2 | - | 2 - 3 | 1564 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Capra hircus Probe spans exons |