Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPDYA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005686857.3 | XP_005686914.1 | 1 - 2 | 565 | 101 | |
XM_013967794.2 | XP_013823248.1 | 2 - 3 | 416 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |