Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FOXN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005691687.2 | XP_005691744.2 | 7 - 8 | 905 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |