Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018063840.1 | XP_017919329.1 | 3 - 4 | 322 | 111 | |
XM_018063841.1 | XP_017919330.1 | 3 - 4 | 230 | 111 | ||
GenBank mRNA | JO577132.1 | - | 3 - 4 | 642 | 111 | |
JO577133.1 | - | 3 - 4 | 642 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |