Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005696786.3 | XP_005696843.1 | 7 - 8 | 1039 | 75 | |
GenBank mRNA | JO433239.2 | - | 3 - 4 | 516 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |