Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005696786.3 | XP_005696843.1 | 8 - 9 | 1138 | 110 | |
GenBank mRNA | JO433239.2 | - | 2 - 3 | 417 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |