Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB33A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018045195.1 | XP_017900684.1 | 850 | 68 | ||
GenBank mRNA | JO637369.2 | - | 639 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus |