Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IKZF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013963958.2 | XP_013819412.1 | 2 - 3 | 156 | 73 | |
XM_018048092.1 | XP_017903581.1 | 2 - 3 | 1440 | 73 | ||
XM_018048093.1 | XP_017903582.1 | 3 - 4 | 1534 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |