Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METRNL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018065384.1 | XP_017920873.1 | 2 - 3 | 779 | 94 | |
GenBank mRNA | KA313773.1 | - | 2 - 3 | 876 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Capra hircus Probe spans exons |