Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013973561.2 | XP_013829015.2 | 7 - 8 | 1099 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |