Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIFM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018042503.1 | XP_017897992.1 | 4 - 5 | 512 | 64 | |
XM_018042504.1 | XP_017897993.1 | 5 - 6 | 608 | 64 | ||
XM_018042506.1 | XP_017897995.1 | 4 - 5 | 508 | 64 | ||
XM_018042507.1 | XP_017897996.1 | 4 - 5 | 512 | 64 | ||
XM_018042508.1 | XP_017897997.1 | 4 - 5 | 512 | 64 | ||
GenBank mRNA | JO634274.2 | - | 4 - 5 | 541 | 64 | |
KA325308.1 | - | 6 - 7 | 1041 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Capra hircus Probe spans exons |