Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018042863.1 | XP_017898352.1 | 2 - 3 | 2090 | 90 | |
GenBank mRNA | KA321491.1 | - | 2 - 3 | 753 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |