Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C6H4orf17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018049278.1 | XP_017904767.1 | 8 - 9 | 1022 | 101 | |
XM_018049279.1 | XP_017904768.1 | 7 - 8 | 987 | 101 | ||
XM_018049280.1 | XP_017904769.1 | 7 - 8 | 945 | 101 | ||
XM_018049281.1 | XP_017904770.1 | 7 - 8 | 692 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |