Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPEB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018049737.1 | XP_017905226.1 | 2 - 3 | 1965 | 76 | |
XM_018049738.1 | XP_017905227.1 | 2 - 3 | 1965 | 76 | ||
XM_018049739.1 | XP_017905228.1 | 2 - 3 | 2299 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |