Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005683835.3 | XP_005683892.1 | 1 - 2 | 344 | 58 | |
XM_018052140.1 | XP_017907629.1 | 2 - 3 | 792 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |