Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: N4BP2L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005687525.3 | XP_005687582.2 | 8 - 9 | 3707 | 116 | |
XM_013968170.2 | XP_013823624.1 | 7 - 8 | 2340 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |