Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018058926.1 | XP_017914415.1 | 5 - 6 | 866 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons |