Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: R3HCC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018052332.1 | XP_017907821.1 | 7 - 8 | 1397 | 66 | |
GenBank mRNA | JO396282.2 | - | 1 - 2 | 290 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Capra hircus Probe spans exons |