Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEFM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013966130.2 | XP_013821584.2 | 1 - 2 | 1753 | 66 | |
GenBank mRNA | JO634510.2 | - | 1 - 2 | 1230 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons |