Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFSD9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018054965.1 | XP_017910454.1 | 3 - 4 | 363 | 72 | |
GenBank mRNA | JO439582.2 | - | 3 - 4 | 822 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Capra hircus Probe spans exons |