Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLRN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005675446.2 | XP_005675503.1 | 1 - 2 | 345 | 69 | |
XM_005675447.2 | XP_005675504.1 | 1 - 2 | 345 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |