Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLRN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005675446.2 | XP_005675503.1 | 2 - 3 | 528 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Capra hircus Probe spans exons |