Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677786.3 | XP_005677843.1 | 7 - 8 | 1107 | 99 | |
XM_005677789.3 | XP_005677846.1 | 7 - 8 | 1125 | 99 | ||
XM_013962496.2 | XP_013817950.1 | 8 - 9 | 1209 | 99 | ||
XM_013962497.2 | XP_013817951.1 | 9 - 10 | 1240 | 99 | ||
GenBank mRNA | JO420799.2 | - | 1 - 2 | 1338 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |