Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EEF1AKMT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005680263.3 | XP_005680320.1 | 2 - 3 | 339 | 93 | |
GenBank mRNA | JO429299.2 | - | 2 - 3 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |