Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OCEL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005682133.3 | XP_005682190.1 | 3 - 4 | 748 | 85 | |
XM_018051368.1 | XP_017906857.1 | 4 - 5 | 828 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Capra hircus Probe spans exons |