Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018053382.1 | XP_017908871.1 | 4 - 5 | 1178 | 83 | |
GenBank mRNA | JO589218.2 | - | 1 - 2 | 291 | 83 | |
JO589219.2 | - | 1 - 2 | 291 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Capra hircus Probe spans exons |