Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLDN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018054194.1 | XP_017909683.1 | 9 - 10 | 1191 | 68 | |
XM_018054196.1 | XP_017909685.1 | 8 - 9 | 1173 | 68 | ||
GenBank mRNA | JO426669.1 | - | 1 - 2 | 1810 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Capra hircus Probe spans exons |