Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005692224.3 | XP_005692281.1 | 5 - 6 | 697 | 110 |
Gene Symbol: LOC108635065 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018045364.1 | XP_017900853.1 | 1 - 2 | 108 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Capra hircus Probe spans exons |