Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018042543.1 | XP_017898032.1 | 2 - 3 | 321 | 112 | |
GenBank mRNA | JO447546.2 | - | 2 - 3 | 464 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |