Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRRT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018038960.1 | XP_017894449.1 | 2 - 3 | 1007 | 77 | |
XM_018038961.1 | XP_017894450.1 | 3 - 4 | 854 | 77 | ||
GenBank mRNA | JO646168.2 | - | 1 - 2 | 184 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons |