Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CATSPER3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018050360.1 | XP_017905849.1 | 1 - 2 | 152 | 125 | |
XM_018050361.1 | XP_017905850.1 | 1 - 2 | 152 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |