Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXB5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018064933.1 | XP_017920422.1 | 1 - 2 | 1792 | 66 | |
GenBank mRNA | JO636599.2 | - | 1 - 2 | 137 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons |