Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PI16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018038971.1 | XP_017894460.1 | 6 - 7 | 1472 | 88 | |
XM_018038972.1 | XP_017894461.1 | 6 - 7 | 1343 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Capra hircus Probe spans exons |