Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018042569.1 | XP_017898058.1 | 2 - 3 | 1240 | 108 | |
GenBank mRNA | JO631800.2 | - | 1 - 2 | 937 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Capra hircus Probe spans exons |