Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSTA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005675022.3 | XP_005675079.1 | 1 - 2 | 120 | 120 | |
GenBank mRNA | JT737688.1 | - | 1 - 2 | 95 | 120 | |
KA307060.1 | - | 2 - 3 | 365 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Capra hircus Probe spans exons |